2-Iodoaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

12230-0050 12230-0250 12230-1000
TS12230-0050EA 53 USD
TS12230-0050 TS12230-0250 TS12230-1000
2-Iodoaniline 98%
2-Iodoaniline
Formula: C₆H₆IN
MW: 219.02 g/mol
Melting Pt: 53…58 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0

Order Now

Specification Test Results

Appearance White to yellow to beige to brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 53 to 58 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR