(4-Hydroxyphenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxyphenylacetic Acid

12171-0250 12171-1000
TS12171-0250EA 45 USD
TS12171-0250 TS12171-1000
(4-Hydroxyphenyl)acetic acid 98%
(4-Hydroxyphenyl)acetic acid
Formula: (C₆H₄(OH))CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004347
CAS Number: 156-38-7
EINECS: 205-851-3

Order Now

Specification Test Results

Appearance White to cream to tan Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 152 °C
Water ≤0.5 % (K.F.)
TLC ≤1 % Methyl ester derivative
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR