(3-Hydroxyphenyl)acetic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

12170-0050 12170-0250
TS12170-0050EA 66 USD
TS12170-0050 TS12170-0250
(3-Hydroxyphenyl)acetic acid 99+%
(3-Hydroxyphenyl)acetic acid
Formula: (C₆H₄(OH))CH₂COOH
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00004337
CAS Number: 621-37-4
EINECS: 210-684-4

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 131 °C
HPLC ≥99.0 %
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR