(2-Hydroxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxyphenylacetic acid

12169-0100 12169-0250
TS12169-0100EA 50 USD
TS12169-0100 TS12169-0250
(2-Hydroxyphenyl)acetic acid 99%
(2-Hydroxyphenyl)acetic acid
Formula: (C₆H₄OH)CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 328 °C (1013 hPa)
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00004323
CAS Number: 614-75-5
EINECS: 210-393-2

Order Now

Specification Test Results

Appearance Light pink to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 148 °C
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR