2-Hydroxy-1-naphthaldehyde tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
12160-1000
12160-5000
TS12160-1000EA
63
USD
TS12160-1000
TS12160-5000
2-Hydroxy-1-naphthaldehyde tech.
2-Hydroxy-1-naphthaldehyde
Formula:
C₁₁H₈O₂ Boiling Pt: 192 °C (27 mmHg) Melting Pt: >77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004005 CAS Number: 708-06-5 EINECS: 211-902-0 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Yellow to brown Moist powder |
Infrared spectrum | Conforms |
Melting point | ≥77 °C (on dry basis) |
Water | 25 to 60 % (K.F.) |
HPLC | ≥93 % (on dry basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...