Oxybenzone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone

12136-0050 12136-1000
TS12136-0050EA 32 USD
TS12136-0050 TS12136-1000
Oxybenzone 98%
Oxybenzone
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 150…160 °C (5 mmHg)
Melting Pt: >62.5 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008387
CAS Number: 131-57-7
EINECS: 205-031-5
Merck Index: 12,07088

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point ≥62.0 °C
GC ≥97.5 %
Specific extinction E (1%/1cm) ≥400 (321 to 327 nm, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR