Vanillic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3-methoxybenzoic acid

12135-0250 12135-1000
TS12135-0250EA 46 USD
TS12135-0250 TS12135-1000
Vanillic acid 97+%
Vanillic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 353
Melting Pt: 210…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8
Merck Index: 12,10068

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 208 to 214 °C
HPLC ≥97.0 %
GC ≥99.0 %
Color scale ≤50 APHA
Free acid ≤0.5 % (as Crotonic acid)
Suitability (for use with Dithizone) Passes test
Substance darkened by H2SO4 ≤5 APHA
Free chlorine ≤5 ppm
Calcium (Ca) ≤0.1 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.2 ppm
Zinc (Zn) ≤0.05 ppm
Impurity ≤0.005 % Trichloroethylene
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR