3-Hydroxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Hydroxyphenyl)methanol, KSD 2405, 3-(Hydroxymethyl)phenol

12102-0050 12102-0250 12102-5000
TS12102-0050EA 36 USD
TS12102-0050 TS12102-0250 TS12102-5000
3-Hydroxybenzyl alcohol 97%
3-Hydroxybenzyl alcohol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004643
CAS Number: 620-24-6
EINECS: 210-633-6

Order Now

Specification Test Results

Appearance White to beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 71 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR