4-Hydroxybenzoic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Hydroxybenzoic Acid

12099-0010 12099-1000 12099-5000
TS12099-0010EA 87 USD
TS12099-0010 TS12099-1000 TS12099-5000
4-Hydroxybenzoic acid 99+%
4-Hydroxybenzoic acid
Formula: C₇H₆O₃
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 214…217 °C
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00002547
CAS Number: 99-96-7
EINECS: 202-804-9
Merck Index: 13,04837

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 214 to 217 °C
HPLC ≥99.0 %
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR