(±)-2-Heptanol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol

12037-0010 12037-0500 12037-2500
TS12037-0010EA 226 USD
TS12037-0010 TS12037-0500 TS12037-2500
(±)-2-Heptanol 99+%
(±)-2-Heptanol
Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 160…162 °C (760 mmHg)
Density: 0.8175 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00004587
CAS Number: 543-49-7
EINECS: 208-844-3
UN: 1987
ADR: 3,III
Merck Index: 13,04682

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥99.0 %
Specific gravity (20°C) 0.816 to 0.819

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR