2-Fluorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11966-0025
11966-1000
11966-5000
TS11966-0025EA
450
USD
TS11966-0025
TS11966-1000
TS11966-5000
2-Fluorotoluene 99%
2-Fluorotoluene
Formula:
C₇H₇F Boiling Pt: 113…114 °C (760 mmHg) Melting Pt: –62 °C Density: 1 g/cm³ Flash Pt: 8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000322 CAS Number: 95-52-3 EINECS: 202-428-5 UN: 2388 ADR: 3,II Merck Index: 13,04207 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4720 to 1.4750 (20°C, 589 nm) |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Specific optical rotation | -9.5° to -11.5° (20°C, 589 nm) (c=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...