4-Fluorophenyl isocyanate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene

11960-0050 11960-0250
TS11960-0050EA 59 USD
TS11960-0050 TS11960-0250
4-Fluorophenyl isocyanate 99%
4-Fluorophenyl isocyanate
Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 42…47 °C (1 mmHg)
Density: 1.206 g/cm³
Flash Pt: 60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002023
CAS Number: 1195-45-5
EINECS: 214-799-0
UN: 3080
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5130 to 1.5150 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR