4-Fluorophenylhydrazinium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride

11959-0100 11959-0250 11959-1000
TS11959-0100EA 64 USD
TS11959-0100 TS11959-0250 TS11959-1000
4-Fluorophenylhydrazinium chloride 97%
4-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: 275…277 °C
Storage Temperature: Ambient
MDL Number: MFCD00012942
CAS Number: 823-85-8
EINECS: 212-521-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
GC ≥96 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR