(4-Fluorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Fluorophenyl)acetic acid

11957-0100 11957-0250 11957-1000
TS11957-0100EA 41 USD
TS11957-0100 TS11957-0250 TS11957-1000
(4-Fluorophenyl)acetic acid 98%
(4-Fluorophenyl)acetic acid
Formula: (C₆H₄F)CH₂COOH
Melting Pt: 85…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00004343
CAS Number: 405-50-5
EINECS: 206-972-4
Merck Index: 13,04204

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 81 to 85 °C
HPLC ≥97.5 %
Stabilizer 0.1 % w/w hydroquinone


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR