4-Fluorophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene

11956-0250 11956-1000
TS11956-0250EA 42 USD
TS11956-0250 TS11956-1000
4-Fluorophenol 99%
4-Fluorophenol
Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 44…49 °C
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002316
CAS Number: 371-41-5
EINECS: 206-736-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to beige to pink Crystalline solid
Infrared spectrum Conforms
Melting point 44 to 49 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR