3-Fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

11955-0100 11955-0500 11955-2000
TS11955-0100EA 53 USD
TS11955-0100 TS11955-0500 TS11955-2000
3-Fluorophenol 98%
3-Fluorophenol
Formula: C₆H₅FO
Boiling Pt: 178 °C (760 mmHg)
Melting Pt: 10 °C
Density: 1.24 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002254
CAS Number: 372-20-3
EINECS: 206-748-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5125 to 1.5155 (20°C, 589 nm)
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR