2-Fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Fluorophenol

11954-0100 11954-1000 11954-5000
TS11954-0100EA 37 USD
TS11954-0100 TS11954-1000 TS11954-5000
2-Fluorophenol 98%
2-Fluorophenol
Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 152 °C (760 mmHg)
Melting Pt: 16.1 °C
Density: 1.256 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00002155
CAS Number: 367-12-4
EINECS: 206-681-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5134 to 1.5154 (20°C, 589 nm)
GC ≥97.5 %
UV at 250 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤2 ppm
Acidity ≤0.0003 meq/g
Filtered to 0.2 micrometer Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR