3-Fluorobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

11937-0050 11937-0250 11937-1000
TS11937-0050EA 22 USD
TS11937-0050 TS11937-0250 TS11937-1000
3-Fluorobenzoic acid 99%
3-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9
EINECS: 207-248-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 121 to 125 °C
GC ≥98.5 %
Titration with TBAH ≥98.5 %
Solubility (in water) Sparingly soluble
Related substances Passes test
Titration with NaOH 99.5 to 100.5 % (On dried substance)
Chloride (Cl) ≤200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR