3-Fluorobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

11933-0100 11933-0500 11933-2500
TS11933-0100EA 43 USD
TS11933-0100 TS11933-0500 TS11933-2500
3-Fluorobenzaldehyde 98+%
3-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 66…68 °C (20 mmHg)
Density: 1.17 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5190 (20°C, 589 nm)
GC ≥98.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR