2-Fluorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

11932-0250 11932-1000
TS11932-0250EA 43 USD
TS11932-0250 TS11932-1000
2-Fluorobenzaldehyde 97%
2-Fluorobenzaldehyde
Formula: C₇H₅FO
Boiling Pt: 172…174 °C
Melting Pt: –44.5 °C
Density: 1.17 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00003302
CAS Number: 446-52-6
EINECS: 207-171-2
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5205 to 1.5225 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR