4-Fluoroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

11929-0250 11929-1000 11929-5000
TS11929-0250EA 25 USD
TS11929-0250 TS11929-1000 TS11929-5000
4-Fluoroaniline 98%
4-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: –1.9 °C
Density: 1.157 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,04196

Order Now

Specification Test Results

Appearance Clear light yellow to red-brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5385 to 1.5405 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR