2-Fluoroaniline 99+%

Supplier: Thermo Scientific Chemicals
Warning

11927-0250 11927-1000 11927-5000
TS11927-0250EA 29 USD
TS11927-0250 TS11927-1000 TS11927-5000
2-Fluoroaniline 99+%
2-Fluoroaniline
Formula: C₆H₆FN
Boiling Pt: 182…183 °C (760 mmHg)
Melting Pt: –29 °C
Density: 1.151 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00007642
CAS Number: 348-54-9
EINECS: 206-478-9
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5430 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR