9-Fluorenone 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9H-Fluoren-9-one

11920-0025 11920-1000 11920-5000
TS11920-0025EA 731 USD
TS11920-0025 TS11920-1000 TS11920-5000
9-Fluorenone 99+%
9-Fluorenone
Formula: C₁₃H₈O
Boiling Pt: 342 °C (760 mmHg)
Melting Pt: 81…85 °C
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00001141
CAS Number: 486-25-9
EINECS: 207-630-7
UN: 3077

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 81 to 85 °C
GC ≥99.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR