4-Ethylresorcinol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Ethylbenzene-1,3-diol
11895-0050
11895-0250
TS11895-0050EA
46
USD
TS11895-0050
TS11895-0250
4-Ethylresorcinol 98%
4-Ethylresorcinol
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 283 °C (1013 hPa) Melting Pt: 95…99 °C |
MDL Number:
MFCD00002283 CAS Number: 2896-60-8 EINECS: 220-777-1 |
Specification Test Results
Appearance | Light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 95 to 99 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...