4-Ethylphenol 97%

Supplier: Thermo Scientific Chemicals
Danger

11882-0010 11882-0050 11882-2500
TS11882-0010EA 198 USD
TS11882-0010 TS11882-0050 TS11882-2500
4-Ethylphenol 97%
4-Ethylphenol
Formula: C₈H₁₀O
Boiling Pt: 218…219 °C (760 mmHg)
Melting Pt: 41…46 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002393
CAS Number: 123-07-9
EINECS: 204-598-6
UN: 2430
ADR: 8,III

Order Now

Specification Test Results

Appearance White to cream to brown Crystals or crystalline solid or fused mass
Infrared spectrum Conforms
Melting point 38 to 45 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR