1,12-Dodecanediol 98%

Supplier: Thermo Scientific Chemicals

11761-0010 11761-0250 11761-1000
TS11761-0010EA 244 USD
TS11761-0010 TS11761-0250 TS11761-1000
1,12-Dodecanediol 98%
1,12-Dodecanediol
Formula: C₁₂H₂₆O₂
Boiling Pt: 324 °C (760 mmHg)
Melting Pt: 79…81 °C
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00004755
CAS Number: 5675-51-4
EINECS: 227-133-9

Order Now

Specification Test Results

Appearance White Crystals or flakes
Infrared spectrum Conforms
Melting point 79 to 81 °C
GC ≥97.5 %
Residue after reduction ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR