3,3-Diphenylpropionic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

11742-0250 11742-1000
TS11742-0250EA 49 USD
TS11742-0250 TS11742-1000
3,3-Diphenylpropionic acid 97%
3,3-Diphenylpropionic acid
Formula: C₁₅H₁₄O₂
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 150…155 °C
Flash Pt: 113 °C
MDL Number: MFCD00002717
CAS Number: 606-83-7
EINECS: 210-125-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 155 °C
GC ≥96.0 %
Titration with NaOH 96.0 to 104.0 %
Iron (Fe) ≤10 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR