Diphenylmethane 99%

Supplier: Thermo Scientific Chemicals
Warning

11735-0010 11735-5000
TS11735-0010EA 107 USD
TS11735-0010 TS11735-5000
Diphenylmethane 99%
Diphenylmethane
Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 24.5…25 °C
Density: 1 g/cm³
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00004781
CAS Number: 101-81-5
EINECS: 202-978-6
UN: 3077
ADR: 9,III
Merck Index: 13,03360

Order Now

Specification Test Results

Appearance Colorless to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥98.5 %
Chlorine and Bromine (as Cl) ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR