trans,trans-1,4-Diphenyl-1,3-butadiene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1E,3E)-1,4-Diphenylbuta-1,3-diene

11722-0250 11722-1000
TS11722-0250EA 104 USD
TS11722-0250 TS11722-1000
trans,trans-1,4-Diphenyl-1,3-butadiene 99%
trans,trans-1,4-Diphenyl-1,3-butadiene
Formula: C₁₆H₁₄
MW: 206.29 g/mol
Melting Pt: 151…154 °C
MDL Number: MFCD00004791
CAS Number: 538-81-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 151 to 154 °C
GC ≥98.5 %
Freezing point 2.0 to 3.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR