9,10-Diphenylanthracene 98%

Supplier: Thermo Scientific Chemicals

11720-0010 11720-0050 11720-0250
TS11720-0010EA 59 USD
TS11720-0010 TS11720-0050 TS11720-0250
9,10-Diphenylanthracene 98%
9,10-Diphenylanthracene
Formula: C₂₆H₁₈
MW: 330.43 g/mol
Melting Pt: 248…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00001253
CAS Number: 1499-10-1
EINECS: 216-105-1

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 248 to 252 °C
HPLC ≥97.5 %
Copper (Cu) ≤0.003 %
Magnesium (Mg) ≤0.002 %
Manganese (Mn) ≤0.01 %
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.003 %
Ferric iron (Fe 3+) ≤100 ppm (May increase due to inherent oxidation on storage)
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR