3,5-Dinitroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11694-0010
11694-0100
TS11694-0010EA
36
USD
TS11694-0010
TS11694-0100
3,5-Dinitroaniline 98%
3,5-Dinitroaniline
Formula:
C₆H₅N₃O₄ Melting Pt: 159…163 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007263 CAS Number: 618-87-1 EINECS: 210-567-8 UN: 1596 ADR: 6.1,II |
Specification Test Results
Appearance | Yellow to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 159 to 163 °C |
HPLC | ≥97.5 % |
Calcium (Ca) | ≤50 ppm |
Copper (Cu) | ≤20 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤20 ppm |
Magnesium (Mg) | ≤50 ppm |
Nickel (Ni) | ≤1500 ppm |
Potassium (K) | ≤100 ppm |
Sodium (Na) | ≤500 ppm |
Zinc (Zn) | ≤100 ppm |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...