N,N-Dimethyl-p-toluidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene

11690-0025 11690-5000
TS11690-0025EA 531 USD
TS11690-0025 TS11690-5000
N,N-Dimethyl-p-toluidine 99%
N,N-Dimethyl-p-toluidine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 211 °C (760 mmHg)
Density: 0.93 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008316
CAS Number: 99-97-8
EINECS: 202-805-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Color scale ≤10 APHA
Stabilizer 0.5 to 1.0 % Ethanol
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Free chlorine (Cl) Passes test
Lead (Pb) ≤0.05 ppm
Acid and chloride Passes test
Acetone and aldehyde Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR