4,5-Dimethyl-o-phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,5-Dimethylbenzene-1,2-diamine, 4,5-Dimethyl-1,2-phenylenediamine, 4,5-Diamino-1,2-dimethylbenzene, 1,2-Diamino-4,5-dimethylbenzene

11661-0050 11661-0250
TS11661-0050EA 85 USD
TS11661-0050 TS11661-0250
4,5-Dimethyl-o-phenylenediamine 98%
4,5-Dimethyl-o-phenylenediamine
Formula: C₈H₁₂N₂
Melting Pt: 127.2…128.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00007729
CAS Number: 3171-45-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to orange to brown Powder or solid
Infrared spectrum Conforms
GC ≥97.5 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.03 %
Sodium (Na) ≤0.05 %
Strontium (Sr) ≤0.2 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %
Oxidizing substance (as NO3) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR