3,4-Xylenol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11658-0025
11658-1000
11658-5000
TS11658-0025EA
374
USD
TS11658-0025
TS11658-1000
TS11658-5000
3,4-Xylenol 99%
3,4-Xylenol
Formula:
C₈H₁₀O Boiling Pt: 227 °C (760 mmHg) Melting Pt: 63…67 °C Density: 0.983 g/cm³ Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002304 CAS Number: 95-65-8 EINECS: 202-439-5 UN: 2928 ADR: 6.1,II Merck Index: 12,10215 |
Specification Test Results
Appearance | White to yellow to brown Crystals or crystalline mass and/or chunks |
Infrared spectrum | Conforms |
Melting point | 63 to 67 °C |
GC | ≥98.5 % |
Color scale | ≤250 APHA |
Residue after ignition | ≤0.005 % |
Hydrocarbons | Passes test |
Chlorobenzene (C6H5Cl) | ≤0.01 % |
Nitrobenzene (C6H5NO2) | (Limit about 0.001 %) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...