2,6-Xylidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-m-xylene, 2,6-Dimethylaniline

11594-0025 11594-0050 11594-1000 11594-5000
TS11594-0025EA 310 USD
TS11594-0025 TS11594-0050 TS11594-1000 TS11594-5000
2,6-Xylidine 99%
2,6-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 216 °C (760 mmHg)
Melting Pt: 10…12 °C
Density: 0.98 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007747
CAS Number: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5591 to 1.5611 (20°C, 589 nm)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR