(3,4-Dimethoxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Homoveratric acid, 3,4-Dimethoxyphenylacetic acid

11561-0250 11561-1000 11561-5000
TS11561-0250EA 34 USD
TS11561-0250 TS11561-1000 TS11561-5000
(3,4-Dimethoxyphenyl)acetic acid 99%
(3,4-Dimethoxyphenyl)acetic acid
Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00004335
CAS Number: 93-40-3
EINECS: 202-244-5

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
Water ≤1 % (K.F.)
Titration with TBAH ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR