2,6-Dimethoxyphenol 99%

Supplier: Thermo Scientific Chemicals
Warning

11559-0250 11559-1000 11559-5000
TS11559-0250EA 52 USD
TS11559-0250 TS11559-1000 TS11559-5000
2,6-Dimethoxyphenol 99%
2,6-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 261 °C (760 mmHg)
Melting Pt: 50…56 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00064434
CAS Number: 91-10-1
EINECS: 202-041-1

Order Now

Specification Test Results

Appearance White to light yellow to grey to brown Crystalline powder or crystals or flakes or fused solid
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR