2,7-Dihydroxynaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,7-Naphthalenediol, Naphthalene-2,7-diol

11504-0050 11504-0250 11504-1000 11504-5000
TS11504-0050EA 26 USD
TS11504-0050 TS11504-0250 TS11504-1000 TS11504-5000
2,7-Dihydroxynaphthalene 97%
2,7-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Melting Pt: 184…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00004085
CAS Number: 582-17-2
EINECS: 209-478-7

Order Now

Specification Test Results

Appearance Grey Powder
Infrared spectrum Conforms
Melting point 184 to 189 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR