2,3-Dihydroxynaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Naphthalene-2,3-diol, 2,3-Naphthalenediol

11502-0500 11502-2500
TS11502-0500EA 64 USD
TS11502-0500 TS11502-2500
2,3-Dihydroxynaphthalene 97%
2,3-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 162…166 °C
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00004073
CAS Number: 92-44-4
EINECS: 202-156-7

Order Now

Specification Test Results

Appearance White to beige to pink to light grey Powder or crystals
Infrared spectrum Conforms
Melting point 161 to 165 °C
HPLC ≥96.0 %
Clarity of solution (c=20, water) Passes test
Sulfate (SO4) ≤0.3 %
Iron (Fe) ≤0.01 %
Sodium (Na) ≤0.002 %
Heavy metals (as Pb) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR