1,5-Dihydroxynaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,5-Naphthalenediol, Naphthalene-1,5-diol

11500-1000 11500-5000
TS11500-1000EA 48 USD
TS11500-1000 TS11500-5000
1,5-Dihydroxynaphthalene 97%
1,5-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 259…261 °C (dec.)
Flash Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00003980
CAS Number: 83-56-7
EINECS: 201-487-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Grey-brown to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR