Caffeic acid predominantly trans 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

11493-0050 11493-0250 11493-1000
TS11493-0050EA 50 USD
TS11493-0050 TS11493-0250 TS11493-1000
Caffeic acid predominantly trans 99+%
Caffeic acid
Formula: C₉H₈O₄
Melting Pt: 194…198 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004392
CAS Number: 331-39-5
EINECS: 206-361-2
Merck Index: 12,01673

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Titration with NaOH ≥99.0 %
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR