4,4'-Dihydroxybenzophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(4-hydroxyphenyl) ketone

11492-0050 11492-0250
TS11492-0050EA 56 USD
TS11492-0050 TS11492-0250
4,4'-Dihydroxybenzophenone 97%
4,4'-Dihydroxybenzophenone
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 217…222 °C
MDL Number: MFCD00002358
CAS Number: 611-99-4
EINECS: 210-288-1

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 217 to 222 °C
GC ≥96.0 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR