3,5-Dihydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-Resorcylic acid

11490-1000 11490-5000
TS11490-1000EA 56 USD
TS11490-1000 TS11490-5000
3,5-Dihydroxybenzoic acid 97%
3,5-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 235…240 °C
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002512
CAS Number: 99-10-5
EINECS: 202-730-7

Order Now

Specification Test Results

Appearance White to beige to light pink Powder
Infrared spectrum Conforms
Melting point 235 to 242 °C
HPLC ≥96.0 %
Titration with NaOH 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR