2,6-Dihydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

11488-0250 11488-1000 11488-5000
TS11488-0250EA 51 USD
TS11488-0250 TS11488-1000 TS11488-5000
2,6-Dihydroxybenzoic acid 97%
2,6-Dihydroxybenzoic acid
Formula: C₇H₆O₄
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002462
CAS Number: 303-07-1
EINECS: 206-134-8

Order Now

Specification Test Results

Appearance White to light yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 160 to 165 °C (Dec)
GC ≥96.0 %
Titration with NaOH 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR