2,4-Dihydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: beta-Resorcylic acid

11487-0050 11487-1000 11487-5000
TS11487-0050EA 26 USD
TS11487-0050 TS11487-1000 TS11487-5000
2,4-Dihydroxybenzoic acid 97%
2,4-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 390 °C (1013 hPa)
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00002451
CAS Number: 89-86-1
EINECS: 201-946-9
Merck Index: 12,08325

Order Now

Specification Test Results

Appearance Cream to light beige to pink Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR