1,4-Difluorobenzene 99+%

Supplier: Thermo Scientific Chemicals
Danger

11461-0250 11461-1000
TS11461-0250EA 68 USD
TS11461-0250 TS11461-1000
1,4-Difluorobenzene 99+%
1,4-Difluorobenzene
Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.17 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000344
CAS Number: 540-36-3
EINECS: 208-742-9
UN: 1993
ADR: 3,II
Merck Index: 13,03172

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR