1,3-Difluorobenzene 99+%

Supplier: Thermo Scientific Chemicals
Danger

11460-0100 11460-0250 11460-1000
TS11460-0100EA 61 USD
TS11460-0100 TS11460-0250 TS11460-1000
1,3-Difluorobenzene 99+%
1,3-Difluorobenzene
Formula: C₆H₄F₂
Boiling Pt: 83 °C (760 mmHg)
Melting Pt: –59 °C
Density: 1.15 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000327
CAS Number: 372-18-9
EINECS: 206-746-5
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4390 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR