2,4-Difluoroaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

11459-0100 11459-0250 11459-1000
TS11459-0100EA 35 USD
TS11459-0100 TS11459-0250 TS11459-1000
2,4-Difluoroaniline 99%
2,4-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 170 °C (753 mmHg)
Melting Pt: –7.5 °C
Density: 1.268 g/cm³
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007648
CAS Number: 367-25-9
EINECS: 206-687-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,03171

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5055 to 1.5085 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer ≤1.1 % Triethylamine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR