Diethyl phthalate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Phthalic acid diethyl ester

11452-0010 11452-0025 11452-0100 11452-0250
TS11452-0010EA 63 USD
TS11452-0010 TS11452-0025 TS11452-0100 TS11452-0250
Diethyl phthalate 99%
Diethyl phthalate
Formula: C₁₂H₁₄O₄
Boiling Pt: 298…299 °C (760 mmHg)
Density: 1.118 g/cm³
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=2461e9f0e230ca44b5372d1233410fbf47a564c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5010 to 1.5030 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.5 %
Color scale ≤25 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR