3,5-Dichlorophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

11361-0100 11361-0500
TS11361-0100EA 100 USD
TS11361-0100 TS11361-0500
3,5-Dichlorophenol 99%
3,5-Dichlorophenol
Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 233 °C (760 mmHg)
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002259
CAS Number: 591-35-5
EINECS: 209-714-9
UN: 2020
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder or crystals or crystalline mass
Infrared spectrum Conforms
Melting point 64 to 68 °C
GC ≥98.5 %
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR